missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C14orf80 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558636
This item is not returnable.
View return policy
Description
Immunogen sequence: RVLSPLPAGN ALASLALEVQ ARLVKSALCS QGYPRLALAQ LPEDGSQGSR ELLLALSWLL ARGPVPEQML AQARVPLGDE MTV Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 65%.
C14orf80 may play a role in counteracting perturbation of actin filaments, such as after treatment with the actin depolymerizing microbial metabolite Chivosazole F.
Specifications
| C14orf80 | |
| Polyclonal | |
| Unconjugated | |
| TEDC1 | |
| C14orf80; chromosome 14 open reading frame 80; TEDC1; Tubulin epsilon and delta complex protein 1; uncharacterized protein C14orf80 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86SX3 | |
| TEDC1 | |
| Recombinant protein corresponding to Human C14orf80. Recombinant protein control fragment (Product #RP-102863). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction