missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf39 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | C14orf39 |
|---|---|
| Concentration | 0.34 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16812035
|
Proteintech
22664-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16802035
|
Proteintech
22664-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| C14orf39 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q8N1H7 | |
| 317761, 75801 | |
| C14orf39 Fusion Protein Ag18589 | |
| Primary | |
| -20°C | |
| SIX6OS1 |
| 0.34 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C14orf39, Six6os1 | |
| 4930447C04Rik, SIX6OS1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title