missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C14orf39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100411
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total C14orf39.
Meiotic protein that localizes to the central element of the synaptonemal complex and is required for chromosome synapsis during meiotic recombination. Required for the appropriate processing of intermediate recombination nodules before crossover formation.
Specifications
| C14orf39 | |
| Polyclonal | |
| Unconjugated | |
| C14orf39 | |
| 4921504I02Rik; 4930447C04Rik; A930035O15Rik; C14orf39; chromosome 14 open reading frame 39; protein SIX6OS1; RIKEN cDNA 4930447C04 gene; Six6 antisense; Six6 opposite strand transcript 1; Six6 opposite strand transcript 1 homolog; Six6as; Six6OS; Six6os1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 317761, 75801 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N1H7, Q9CTN5 | |
| 4930447C04Rik, C14orf39 | |
| A synthesized peptide derived from human SIX6OS1(Accession Q8N1H7), corresponding to amino acid residues E528-S578. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction