missing translation for 'onlineSavingsMsg'
Learn More

C14orf28 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen C14orf28
Concentration 0.29 mg/mL
Applications Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16804665
View Documents
Proteintech
26130-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16894655
View Documents
Proteintech
26130-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C14orf28 is a protein coding gene.
Specifications

Specifications

C14orf28
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human
Q4W4Y0
122525
C14orf28 Fusion Protein Ag23441
Primary
-20°C
C14orf28
0.29 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
c14_5270, C14orf28, DRIP1
C14orf28
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.