missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C14orf28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569796
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
C14orf28 is a protein coding gene.
Specifications
| C14orf28 | |
| Polyclonal | |
| Unconjugated | |
| C14orf28 | |
| c14_5270; C14orf28; chromosome 14 open reading frame 28; dopamine receptor interacting protein 1; dopamine receptor-interacting protein 1; DRIP1; DRIP-1; uncharacterized protein C14orf28 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 122525 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q4W4Y0 | |
| C14orf28 | |
| synthetic peptide directed towards the middle region of human C14orf28. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction