Learn More
C14orf166 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19848-1-AP-150UL
 View more versions of this product
Product Code. 16809334
  • £349.00 / 150µL
Description
The function of the C14orf266 gene has not yet been determined.
Specifications
Specifications
C14orf166
Polyclonal
Unconjugated
C14orf166
C14orf166, CGI 99, CGI99, CLE, CLE7, LCRP369, RLLM1, UPF0568 protein C14orf166
Rabbit
Antigen Affinity Chromatography
RUO
302247, 51637, 68045
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y224
C14orf166, Rtraf
C14orf166 Fusion Protein Ag13900
150 μL
Primary
Human, Mouse, Rat