missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf166 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19848-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of the C14orf266 gene has not yet been determined.Specifications
| C14orf166 | |
| Polyclonal | |
| Unconjugated | |
| C14orf166 | |
| C14orf166, CGI 99, CGI99, CLE, CLE7, LCRP369, RLLM1, UPF0568 protein C14orf166 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 302247, 51637, 68045 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y224 | |
| C14orf166, Rtraf | |
| C14orf166 Fusion Protein Ag13900 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction