missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTRAF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106393
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total C14orf166.
RTRAF (C14orf166) is an RNA-binding protein involved in modulation of mRNA transcription by Polymerase II (PubMed:16950395). Component of the tRNA-splicing ligase complex and is required for tRNA ligation (PubMed:24870230). May be required for RNA transport (PubMed:24608264).
Specifications
| RTRAF | |
| Polyclonal | |
| Unconjugated | |
| RTRAF | |
| 2700060E02Rik; C14orf166; CGI99; CGI-99; chromosome 14 open reading frame 166; CLE; CLE7; CLE7 homolog; hCLE1; LCRP369; LRRGT00192; RGD1304704; RIKEN cDNA 2700060E02 gene; RLL motif containing 1; RLLM1; RNA transcription, translation and transport factor protein; RTRAF; similar to Hypothetical protein CGI-99; UPF0568 protein C14orf166; UPF0568 protein C14orf166 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 302247, 51637, 68045 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CQE8, Q9Y224 | |
| RTRAF | |
| A synthesized peptide derived from human C14orf166(Accession Q9Y224), corresponding to amino acid residues E24-Q74. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction