missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTRAF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540917
This item is not returnable.
View return policy
Description
Peptide sequence: LPVALDKHIL GFDTGDAVLN EAAQILRLLH IEELRELQTK INEAIVAVQA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
RTRAF (C14orf166) is an RNA-binding protein involved in modulation of mRNA transcription by Polymerase II (PubMed:16950395). Component of the tRNA-splicing ligase complex and is required for tRNA ligation (PubMed:24870230). May be required for RNA transport (PubMed:24608264).
Specifications
| RTRAF | |
| Polyclonal | |
| Unconjugated | |
| RTRAF | |
| 2700060E02Rik; C14orf166; CGI99; CGI-99; chromosome 14 open reading frame 166; CLE; CLE7; CLE7 homolog; hCLE1; LCRP369; LRRGT00192; RGD1304704; RIKEN cDNA 2700060E02 gene; RLL motif containing 1; RLLM1; RNA transcription, translation and transport factor protein; RTRAF; similar to Hypothetical protein CGI-99; UPF0568 protein C14orf166; UPF0568 protein C14orf166 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51637 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y224 | |
| RTRAF | |
| Synthetic peptide directed towards the middle region of human C14orf166. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction