missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C14orf130 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569234
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway, recognizes and binds to proteins bearing specific amino-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation.
Specifications
| C14orf130 | |
| Polyclonal | |
| Unconjugated | |
| UBR7 | |
| 5730410I19Rik; AA589405; AW557761; C14orf130; C21H14ORF130; fb74a06; hypothetical protein LOC322815; LOW QUALITY PROTEIN: putative E3 ubiquitin-protein ligase UBR7; N-recognin-7; Putative E3 ubiquitin-protein ligase UBR7; RGD1359144; RING-type E3 ubiquitin transferase UBR7; ubiquitin protein ligase E3 component n-recognin 7; ubiquitin protein ligase E3 component n-recognin 7 (putative); Ubr7; wu:fb74a06; wu:fi29b06; zgc:55404 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 314399 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| UBR7 | |
| synthetic peptide directed towards the N-terminal region of rat C14orf130. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction