missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C13ORF39 Mouse anti-Human, Purified MaxPab™ Polyclonal , Abnova™
Mouse polyclonal antibody raised against a full-length human C13orf39 protein.
Brand: Abnova H00196541-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MDVCLSSAQQPGRRGEGLSSPGGWLEAEKKGAPQKDSTGGVLEESNKIEPSLHSLQKFVPTDYASYTQEHYRFAGKEIVIQESIESYGAVVWPGAMALCQYLEEHAEELNFQDAKILEIGAGPGLVSIVASILGAQVTATDLPDVLGNLQYNLLKNTLQCTAHLPEVKELVWGEDLDKNFPKSAFYYDYVLASDVVYHHYFLDKLLTTMVYLSQPGTVLLWANKFRFSTDYEFLDKFKQVFDTTLLAEYPESSVKLFKGILKWDSpecifications
| C13ORF39 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human C13orf39 protein. | |
| METTL21C | |
| C13orf39 | |
| C13orf39 (NP_001010977,1 a.a. ∽ 264 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Antibody reactive against mammalian transfected lysate. | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001010977 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 196541 | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction