missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C12orf53 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA525109

Product Code. 13212689

  • £443.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with non-human primate and rat based on sequence homology.

The function of this protein has not been specifically defined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C12orf53
Polyclonal
Unconjugated
PIANP
AI255183; AU067746; Brain protein 1; C12orf53; C530028O21Rik; LEDA1; leda-1; liver endothelial differentiation-associated protein-1; paired immunoglobin-like type 2 receptor-associating neural protein; PANP; Pianp; PILR alpha associated neural protein; PILR alpha-associated neural protein; PILR-associating neural protein; RGD1304952; UNQ828/PRO1755
Rabbit
Antigen affinity chromatography, Protein A
RUO
196500, 319352
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6P1B3, Q8IYJ0
PIANP
KLH conjugated synthetic peptide between 75-103 amino acids from the N-terminal region of human C12orf53.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Il vostro feedback è stato inviato