missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C12orf40 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15838433
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15838433 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15838433 Supplier Invitrogen™ Supplier No. PA570023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Horse: 83%; Human: 100%; Pig: 83%.

C12orf40 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf40
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C12orf40
Gene Accession No. Q86WS4
Gene Alias C12orf40; chromosome 12 open reading frame 40; epididymis luminal protein 206; epididymis secretory protein Li 94; HEL-206; HEL-S-94; uncharacterized protein C12orf40
Gene Symbols C12orf40
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human C12orf40.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283461
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.