missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C12orf4 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21423MFV500
This item is not returnable.
View return policy
Description
C12orf4 Polyclonal antibody specifically detects C12orf4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| C12orf4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| chromosome 12 open reading frame 4, hypothetical protein LOC57102 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C12orf4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 57102 | |
| Store at 4C in the dark. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur