missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C12orf10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114258
This item is not returnable.
View return policy
Description
Immunogen sequence: PLYTRHRMLG PESVPPPKRS RSKLMAPPRI GTHNGTFHCD EALACALLRL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Horse: 85%; Human: 100%; Rat: 90%.
C12orf10 is a protein coding gene.
Specifications
| C12orf10 | |
| Polyclonal | |
| Unconjugated | |
| C12orf10 | |
| C12orf10; chromosome 12 open reading frame 10; Gamm1; melanocyte proliferating gene 1; melanocyte related; MST024; MSTP024; MYG; MYG1; UPF0160 protein MYG1, mitochondrial | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 60314 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HB07 | |
| C12orf10 | |
| Synthetic peptide directed towards the N-terminal region of Human C12orf10 (aa 18-67). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction