missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf80 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25957-1-AP-150UL
This item is not returnable.
View return policy
Description
C11orf80 locates at the chromosome 11. It has two isoforms with MW 70-75 kDa.Specifications
| C11orf80 | |
| Polyclonal | |
| Unconjugated | |
| TOP6BL | |
| C11orf80 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79703 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N6T0 | |
| TOP6BL | |
| C11orf80 Fusion Protein Ag22895 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction