Learn More
C11orf70 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20780-1-AP-20UL
 View more versions of this product
Product Code. 16810355
  • £135.00 / 20µL
Specifications
C11orf70
Polyclonal
Unconjugated
C11orf70
CK070, C11orf70
Rabbit
Antigen Affinity Chromatography
RUO
85016
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BRQ4
C11orf70
C11orf70 Fusion Protein Ag14793
20 μL
Primary
Mouse, Human