Learn More
C11orf66 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558647
Additional Details : Weight : 0.02000kg
Descrizione
Immunogen sequence: TSSGYGREKP SAGPPTKEVR KVHFDTQEHG PQAITGLEPR EVPLLHQQQG QDPLERENFR HGPRFMTSEY NSKYLRDPLD QPDFLQKKSI GAKEGSGFTK QSHQ Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 68%.
PPP1R32 is a protein coding gene.Specifica
C11orf66 | |
Polyclonal | |
Unconjugated | |
PPP1R32 | |
C11orf66; IIIG9; PPP1R32; Protein phosphatase 1 regulatory subunit 32; protein phosphatase 1, regulatory subunit 32 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z5V6 | |
PPP1R32 | |
Recombinant protein corresponding to Human C11orf66 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only