missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C11orf57 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112094
This item is not returnable.
View return policy
Description
C11orf57 Polyclonal Antibody for Western Blot
C11orf57 has three isoforms with MW 30-34 kDa.
Specifications
| C11orf57 | |
| Polyclonal | |
| Unconjugated | |
| Nkapd1 | |
| AU019823; C11orf57; chromosome 11 open reading frame 57; expressed sequence AU019823; Gm639; NKAP domain containing protein 1; NKAPD1; uncharacterized protein C11orf57; uncharacterized protein C11orf57 homolog; Uncharacterized protein NKAPD1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 55216 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q6ZUT1 | |
| Nkapd1 | |
| A synthetic peptide corresponding to the C-terminus of the Human C11orf57. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction