missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf52 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97392IR
This item is not returnable.
View return policy
Description
C11orf52 Polyclonal specifically detects C11orf52 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| C11orf52 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| chromosome 11 open reading frame 52, FLJ25219, hypothetical protein LOC91894, MGC131888, MGC14839 | |
| Produced in rabbits immunized with E. coli-derived Human C11orf52 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 91894 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction