missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C11orf42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569932
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 79%; Rat: 86%.
C11orf42 is a protein coding gene.
Specifications
| C11orf42 | |
| Polyclonal | |
| Unconjugated | |
| C11orf42 | |
| C11orf42; chromosome 11 open reading frame 42; uncharacterized protein C11orf42 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 160298 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N5U0 | |
| C11orf42 | |
| synthetic peptide directed towards the middle region of human C11orf42 (aa 259-308). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction