Learn More
Invitrogen™ C11orf30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596516
Description
Immunogen sequence: SLMEAQIDTN VEHMIVDPPK KALATSMLTG EAGSLPSTHM VVAGMANSTP QQQKCRESCS SPSTVGSSLT TRKIDPPAVP ATGQFMRIQN VGQKKAEESP AEIIIQAIPQ YAIPCHSSSN VVVEPSGLLE LNNFTSQQLD DEETAMEQDI DSSTEDGTEP SPSQSSAERS; Positive Samples: Mouse lung, Mouse brain, Mouse spleen, Mouse ovary, Mouse kidney; Cellular Location: Nucleus.
EMSY is a regulator which is able to repress transcription, possibly via its interaction with a multiprotein chromatin remodeling complex that modifies the chromatin. Its interaction with BRCA2 suggests that it may play a central role in the DNA repair function of BRCA2. As part of a histone H3-specific methyltransferase complex may mediate ligand-dependent transcriptional activation by nuclear hormone receptors. Diseases associated with EMSY include Sporadic Breast Cancer and Breast Cancer.
Specifications
| C11orf30 | |
| Polyclonal | |
| Unconjugated | |
| EMSY | |
| 2210018M11Rik; 2310016L09Rik; AU019521; AU019557; BRCA2-interacting transcriptional repressor EMSY; C11orf30; EMSY; EMSY BRCA2-interacting transcriptional repressor; EMSY protein; EMSY, BRCA2 interacting transcriptional repressor; EMSY, BRCA2-interacting transcriptional repressor; FLJ90741; GL002; protein EMSY; RGD1310712; RIKEN cDNA 2210018M11 gene; similar to EMSY protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 233545, 56946 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z589, Q8BMB0 | |
| EMSY | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1153-1322 of human C11orf30 (NP_0645782). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.