Learn More
Invitrogen™ C11orf24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139743
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VATTANTSSP MSTRPSPSKH MPSDTAASPV PPMRPQAQGP ISQVSVDQPV VNTTNKSTPM PSNTTPEPAP TPTVVTTTKA QAREPTASPV PVPHTSPIPE MEAMSPTTQP SPMPYTQRAA GPGTSQAPEQ VETEATPGT Highest antigen sequence indentity to the following orthologs: Mouse - 46%, Rat - 49%.
The function remains unknown.
Specifications
| C11orf24 | |
| Polyclonal | |
| Unconjugated | |
| C11orf24 | |
| C11orf24; chromosome 11 open reading frame 24; DM4E3; FP2568; Protein DM4E3; Uncharacterized protein C11orf24; UNQ1872/PRO4315 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96F05 | |
| C11orf24 | |
| Recombinant protein corresponding to Human C11orf24. Recombinant protein control fragment (Product #RP-90466). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.