missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C11orf16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144791
This item is not returnable.
View return policy
Description
Immunogen sequence: WRYWKRNGPE PCPGKPGTRY SNICKEEKDH KQQRAQTVVV GTTKELVSKA THMKPPQTPP GEAEHRKRSQ SLAICQWN Antigen sequence identity: Mouse 27%; Rat 26%.
The function of C11orf16 remains unknown.
Specifications
| C11orf16 | |
| Polyclonal | |
| Unconjugated | |
| C11orf16 | |
| C11orf16; chromosome 11 open reading frame 16; ck016; uncharacterized protein C11orf16 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQ32 | |
| C11orf16 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 386-465. Recombinant protein control fragment (Product #RP-109768). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction