missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Karl Fischer Titration is the specific standard method for water content determination and gives accurate and precise results within minutes.
One Click™ User Interface
- Large color touchscreen (VGA 5.7 in. color TFT, 640 x 480 pixel) with simple-to-understand and fast-to-operate One Click user interface
- All users have their own homescreen with dedicated shortcut buttons (four per user) to directly start specific routine tasks
- User interface in English, German, French, Spanish, Italian, Chinese, Russian, Polish, Korean and Portuguese available
Diaphragmless Cell
- Choice of a current generator cell with or without a diaphragm
- Diaphragmless cell offers more convenience for most samples as there is no catholyte needed
- Due to the unique design, the diaphragmless cell even meets the demands of very precise low water content determinations
Solvent Manager for Safe Handling
- Takes care of filling, draining and exchange of reagents
- It is controlled by the titrator and full security is guaranteed by overflow protector in the waste bottle
Flexible Data Export
- Broad range of options for handling data
- Options available to export as CSV, PDF or XML files
- Devices such as printers, memory sticks or USB barcode readers can be connected via the USB port and are automatically recognized
- Common USB printers (DIN-A4/US letter) or the USB-P25 compact printer for secure and GLP-compliant paper recording
Communication
- Ethernet (TCP/IP): 1
- COM (serial balance connection): 1
- USB host (printer, memory stick): 2
- Integrated Magnetic Stirrer: 1
- Pump/Stirrer: 1
Methods and Series
- KF standard methods: 1
- Max. number of user methods: 5
- Number of method functions: 10
- Number of samples per series: 120
- Autostart: automatic start after sample addition: Yes
- Max. number of calculations/results per method: 3
- Statistics of up to 120 samples including result limits check, outlier test: Yes
- Recalculation of results: Yes
- Data in Online Screen: time, water count, potential, stirrer speed, consumption, drift: Yes
- Data in measured value table: time, water count, potential, consumption, drift: Yes
- Curves in Online Screen and printout: E-C, E-t, C-t, drift-t, water count-t: Yes
Application
- Liquid and gaseous samples
- Resources and method database
Specifications
- Measurement Range: -2000 to +2000mV
- Resolution/Limits of Error: 0.1mV/2mV
- Polarization Current Range: 0 to 24μA
- Polarization Current Resolution/Limits of Error: 0.1μA/1μA
- Ambient Temperature: 5° to 40°C
- Atmospheric Humidity: max. 80% at 31°C
- Drift Measurement: Online, <2μg/min.
- Measurement Range: 10μg to 200mg
- Resolution: 0.1μg
- Repeatability: 0.3% at >1mg water
- Glass Beaker: 200mL
- Dimensions (D x W x H): 34 x 21 x 31.2cm
- Weight: 3.3kg
Certifications
CE, ISO 9001, ISO 14001
Specifications
Specifications
| Certifications/Compliance | CE, ISO 9001, ISO 14001 |
| Detection Range | 0 to 24μA (Polarization Current Range) |
| Display Type | VGA 5.7 in. color TFT, 640 x 480 pixel |
| Type | Coulometric KF Titrator |
| For Use With (Application) | Liquid and gaseous samples; Resources and method database |
| Includes | No Diaphragm |
| Model | C10SX |
| Range (mV) | -2000 to +2000 mV |
| Resolution | 0.1mV/2mV; 0.1μA/1μA; 0.1μg |
| Sample Size | 120 (per series) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?