missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf67 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55273-25ul
This item is not returnable.
View return policy
Description
C10orf67 Polyclonal specifically detects C10orf67 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| C10orf67 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| chromosome 10 open reading frame 67, hypothetical protein LOC256815, MGC46732, RP11-792P23.2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| C10ORF67 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EELDQYKDFGFHKMESFAKETSSPKSNLEKENLEYKVENERLLQIISELEEEIQINLKENSGLEDELISMKEMAEKDHKTIQK | |
| 25 μL | |
| Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers | |
| 256815 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction