missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20420-1-AP-20UL
This item is not returnable.
View return policy
Description
FAM208B is a protein coding gene.Specifications
| C10orf18 | |
| Polyclonal | |
| Unconjugated | |
| FAM208B | |
| C10orf18, KIAA2006 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54906 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5VWN6 | |
| FAM208B | |
| C10orf18 Fusion Protein Ag14240 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu