missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20420-1-AP-150UL
This item is not returnable.
View return policy
Description
FAM208B is a protein coding gene.Specifications
| C10orf18 | |
| Polyclonal | |
| Unconjugated | |
| FAM208B | |
| C10orf18, KIAA2006 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54906 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5VWN6 | |
| FAM208B | |
| C10orf18 Fusion Protein Ag14240 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction