missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1 inhibitor Recombinant Rabbit Monoclonal Antibody (0D3W8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537889
This item is not returnable.
View return policy
Description
Positive Samples: Human plasma, Mouse plasma, Rat plasma Immunogen sequence: SLKLYHAFSA MKKVETNMAF SPFSIASLLT QVLLGAGENT KTNLESILSY PKDFTCVHQA LKGFTTKGVT SVSQIFHSPD LAIRDTFVNA SRTLYSSSPR V.
C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ~104 kDa. Serum concentration has been determined to 180 μg/mL.
Specifications
| C1 inhibitor | |
| Recombinant Monoclonal | |
| 3 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 | |
| P05155, P97290, Q6P734 | |
| SERPING1 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0D3W8 | |
| Unconjugated | |
| SERPING1 | |
| C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1Inh; C1-inhibiting factor; C1nh; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12258, 295703, 710 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction