missing translation for 'onlineSavingsMsg'
Learn More

FGF2 Polyclonal Antibody, Invitrogen™

FGF2 Antibody, PA5-23640, from Invitrogen™. Species Reactivity: Human; Applications: Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot.

Brand:  Invitrogen PA5-23640

Product Code. 13218638

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

FGF2 Antibody, PA5-23640, from Invitrogen. Species Reactivity: Human; Applications: Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot.

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members bind heparin and possess broad mitogenic and angiogenic activities. This protein has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for this gene contains multiple polyadenylation sites, and is alternatively translated from non-AUG (CUG) and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: KLH conjugated synthetic peptide between 163-191 amino acids from the central region of human FGF2
Formulation: 400μL purified antibody (Lot-specific) in PBS and 0.09% sodium azide
Synonyms: BFGF, FGFB, HBGF-2, basic fibroblast growth factor bFGF, heparin-binding growth factor 2, prostatropin
Uniprot Accession: P09038
Entrez GeneID: 2247
Gene Symbol: FGF2
This antibody is predicted to react with bovine, chicken, mouse, rat and rabbit based on sequence homology.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FGF2
Polyclonal
P09038
FGF2
KLH conjugated synthetic peptide between 163-191 amino acids from the central region of human FGF2
RUO
2247
Store at 2°-8°C for up to 6 months. For long term storage store at -20°C
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
BFGF, FGFB, HBGF-2, basic fibroblast growth factor bFGF, heparin-binding growth factor 2, prostatropin
Rabbit
400 μL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.