missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. tetani TeNT Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6535-SP
This item is not returnable.
View return policy
Description
TeNT Light Chain/Tetanus Toxin Monoclonal specifically detects TeNT Light Chain/Tetanus Toxin in C. tetani samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TeNT Light Chain/Tetanus Toxin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
| P04958 | |
| TETX | |
| E. coli-derived recombinant C. tetani TeNT Light Chain Pro2-Gly430 Accession # P04958 | |
| 25 μg | |
| Primary | |
| Detects TeNT Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant TeNT Heavy Chain is observed. | |
| C. tetani | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 604023 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TeNT-LC, Tentoxylysin, Tetanus toxin chain H, Tetanus toxin chain L, Tetanus toxin heavy chain, Tetanus toxin light chain, tetX | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1061100 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction