Learn More
c-SRC Mouse anti-Human, CoraLite 594, Clone: 5E10C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60315-100UL
Description
The c-SRC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human c-SRC. This antibody recognizes human, hamster, mouse antigen. The c-SRC antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| c-SRC | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 6714 | |
| IgG2b |
| Immunofluorescence | |
| 5E10C4 | |
| Mouse | |
| Primary | |
| Human, Hamster, Mouse |