missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ c-Rel Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586204
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA., RELB (604758), NFKB1, and NFKB2. These proteins are related through a highly conserved N-terminal region termed the 'Rel domain, ' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (Belguise and Sonenshein, 2007).
Specifications
| c-Rel | |
| Polyclonal | |
| Unconjugated | |
| REL | |
| BCD541; C-Rel; C-Rel protein; C-Rel proto-oncogene protein; gemin 1; gemin-1; LOW QUALITY PROTEIN: proto-oncogene c-Rel; oncogene REL, avian reticuloendotheliosis; proto-oncogene c-Rel; REL; REL proto-oncogene, NF-kB subunit; reticuloendotheliosis oncogene; SMA; SMA1; SMA2; SMA3; SMA4; SMN; SMN2; SMNT; v-rel avian reticuloendotheliosis viral oncogene homolog; v-rel reticuloendotheliosis viral oncogene homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19696, 305584, 5966 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P15307, Q04864 | |
| REL | |
| Synthetic peptide, corresponding to amino acids 470-520 of Human c-Rel. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction