missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ c-Rel Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568782
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 86%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA., RELB (604758), NFKB1, and NFKB2. These proteins are related through a highly conserved N-terminal region termed the 'Rel domain, ' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (Belguise and Sonenshein, 2007).
Specifications
| c-Rel | |
| Polyclonal | |
| Unconjugated | |
| REL | |
| BCD541; C-Rel; C-Rel protein; C-Rel proto-oncogene protein; gemin 1; gemin-1; LOW QUALITY PROTEIN: proto-oncogene c-Rel; oncogene REL, avian reticuloendotheliosis; proto-oncogene c-Rel; REL; REL proto-oncogene, NF-kB subunit; reticuloendotheliosis oncogene; SMA; SMA1; SMA2; SMA3; SMA4; SMN; SMN2; SMNT; v-rel avian reticuloendotheliosis viral oncogene homolog; v-rel reticuloendotheliosis viral oncogene homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19696, 5966 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P15307, Q04864 | |
| REL | |
| synthetic peptide corresponding to a region of mouse c-Rel. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction