Learn More
Invitrogen™ c-Rel Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120137
Description
Positive test controls include: Raji, U-937, A-431, Mouse spleen, Mouse thymus, Rat thymus, Rat lung. The target is usually found in the following locations: Nucleus. Immunogen sequence: MASGAYNPYI EIIEQPRQRG MRFRYKCEGR SAGSIPGEHS TDNNRTYPSI QIMNYYGKGK VRITLVTKND PYKPHPHDLV GKDCRDGYYE AEFGQERRPL FFQNLGIRCV KKKEVKEAII TRIKAGINPF NVPEKQLNDI EDCDLNVVRL CFQVFLPDEH GNLTTALPPV VSNPIYDNRA PNTAELRICR VNKNCGSVRG GDEIFLLCDK VQKDDIEVRF VLNDWEAKGI FSQADVHRQV AIVFKTPPYC KAITEPVTVK MQLRRPSDQE VSESMDFRYL PDEKDTYGNK AKKQKTTLLF.
The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA., RELB (604758), NFKB1, and NFKB2. These proteins are related through a highly conserved N-terminal region termed the 'Rel domain, ' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (Belguise and Sonenshein, 2007).
Specifications
| c-Rel | |
| Polyclonal | |
| Unconjugated | |
| REL | |
| BCD541; C-Rel; C-Rel protein; C-Rel proto-oncogene protein; gemin 1; gemin-1; LOW QUALITY PROTEIN: proto-oncogene c-Rel; oncogene REL, avian reticuloendotheliosis; proto-oncogene c-Rel; REL; REL proto-oncogene, NF-kB subunit; reticuloendotheliosis oncogene; SMA; SMA1; SMA2; SMA3; SMA4; SMN; SMN2; SMNT; v-rel avian reticuloendotheliosis viral oncogene homolog; v-rel reticuloendotheliosis viral oncogene homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19696, 305584, 5966 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.65 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P15307, Q04864 | |
| REL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human REL (NP_0028991). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.