missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Rel Monoclonal Antibody (1RELAH5), eFluor™ 660, eBioscience™

Rat Monoclonal Antibody

£140.00

Specifications

Antigen c-Rel
Clone 1RELAH5
Concentration 0.2 mg/mL
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15559996
View Documents
Invitrogen™
50-6111-80
25 μg
£140.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The 1RELAH5 monoclonal antibody reacts with mouse c-Rel. c-Rel is a 65 kDa hematopoietic lineage-restricted member of the NF-kB family of transcription factors. In the thymus, c-Rel is upregulated in regulatory T cells (Treg) when compared to other CD4+ thymocytes. c-Rel is believed to play a critical role in thymic Treg (nTreg) development, but is not necessary for TGF beta-mediated T reg (iTreg) differentiation. C-Rel is also involved in proliferation and survival in mature T cells. Applications Reported: This 1RELAH5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 1RELAH5 antibody has been tested by intracellular staining and flow cytometric analysis of mouse thymocytes using the Foxp3 Fixation/Permeabilization Buffers (Product # 00-5521-00) and protocol. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser:...

The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA., RELB (604758), NFKB1, and NFKB2. These proteins are related through a highly conserved N-terminal region termed the 'Rel domain, ' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (Belguise and Sonenshein, 2007).
Specifications

Specifications

c-Rel
0.2 mg/mL
Flow Cytometry
eFluor 660
Rat
RUO
PBS with 0.09% sodium azide; pH 7.2
P15307
19696
Primary
Affinity chromatography
1RELAH5
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG2a κ
Mouse
REL
BCD541; C-Rel; C-Rel protein; C-Rel proto-oncogene protein; gemin 1; gemin-1; LOW QUALITY PROTEIN: proto-oncogene c-Rel; oncogene REL, avian reticuloendotheliosis; proto-oncogene c-Rel; REL; REL proto-oncogene, NF-kB subunit; reticuloendotheliosis oncogene; SMA; SMA1; SMA2; SMA3; SMA4; SMN; SMN2; SMNT; v-rel avian reticuloendotheliosis viral oncogene homolog; v-rel reticuloendotheliosis viral oncogene homolog
REL
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado