missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Rel Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-97834AF532
This item is not returnable.
View return policy
Description
c-Rel Polyclonal antibody specifically detects c-Rel in Human samples. It is validated for Western Blot
Specifications
| c-Rel | |
| Polyclonal | |
| Western Blot | |
| C-Rel, C-Rel proto-oncogene protein, I-Rel, oncogene REL, avian reticuloendotheliosis, proto-oncogene c-Rel, v-rel avian reticuloendotheliosis viral oncogene homolog, v-rel reticuloendotheliosis viral oncogene homolog (avian) | |
| The immunogen recognized by this antibody maps to a region between residue 569 and 619 of human reticuloendotheliosis viral oncogene homolog using the numbering given in entry NP_002899.1 (GeneID 5966). | |
| 0.1 mL | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| 5966 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction