missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse C-Reactive Protein/CRP Antibody, R&D Systems™
Rat Monoclonal Antibody
£448.15
Specifications
Antigen | C-Reactive Protein/CRP |
---|---|
Clone | 267917 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
C-Reactive Protein/CRP | |
LYOPH | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
P14847 | |
1401 | |
Mouse myeloma cell line NS0-derived recombinant mouse C-Reactive Protein Met1-Ser225 Accession # P14847 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse C-Reactive Protein in ELISAs. |
267917 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
CRP | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only