missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress C-Phycocyanin

Product Code. 16452439
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452439 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452439 Supplier MedChemExpress Supplier No. HYD1025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress C-phycocyanin (C-PC) is a protein pigment which is also widely used as an excellent nutrient supplement for human beings.

Specifications

CAS 11016-15-2
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Neuroscience-Neuromodulation
Product Type Reference compound
Quantity 1 mg
Shipping Condition Room temperature
Solubility H2O : 14.29 mg/mL (Need ultrasonic)
Synonym C-PC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.