Learn More
Labcon C-Pette™ and C-Pette™ E Blood Transfer Dispensers
These E dispensers provide an easy way to dispense blood into a test cassette without removing the cap from the collection tube.
Brand: Labcon 7625-540-000-9
Additional Details : Weight : 0.45360kg
Description
Both E dispensers fit all standard blood collection tubes and are ideal for use with many Point-of-Care (POC) analyzers that require the delivery of a sample.
- Minimizes aerosols and reduces the risk of lab-acquired infections
- Safe to use at patient bedside
- Polystyrene USP Class VI
- Stainless Steel or Arylic cannula
- Not Autoclavable
- Made in USA
Specifications
Blood Droplet Dispenser | |
Yes | |
Acrylic Cannula | |
100 Pk. |