missing translation for 'onlineSavingsMsg'
Learn More

C-Peptide Antibody (GT1455), Novus Biologicals™

Product Code. 18348245 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348245 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348245 Supplier Novus Biologicals Supplier No. NBP313506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C-Peptide Monoclonal antibody specifically detects C-Peptide in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen C-Peptide
Applications ELISA
Classification Monoclonal
Clone GT1455
Conjugate Unconjugated
Dilution ELISA 1:1000-1:10000
Formulation PBS
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human C-Peptide. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.