missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Peptide Antibody (CC34), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05433IR
This item is not returnable.
View return policy
Description
C-Peptide Monoclonal specifically detects C-Peptide in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA.
Specifications
| C-Peptide | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| INS | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein. | |
| 0.2 mL | |
| Diabetes Research | |
| 3630 | |
| Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| CC34 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reacts with rat c-peptide I, rat c-peptide II, mouse c-peptide II and mouse c-peptide I. Does not react with rat proinsulin. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction