missing translation for 'onlineSavingsMsg'
Learn More

C-Peptide Antibody (CC27), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18416505 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416505 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416505 Supplier Novus Biologicals Supplier No. NBP105434AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C-Peptide Monoclonal antibody specifically detects C-Peptide in Rat samples. It is validated for ELISA, Immunoassay, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen C-Peptide
Applications ELISA, Immunoassay, Sandwich ELISA
Classification Monoclonal
Clone CC27
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.