missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Peptide Antibody (7E10), Novus Biologicals™
Mouse Monoclonal Antibody
£158.00
Specifications
| Antigen | C-Peptide |
|---|---|
| Clone | 70000000000 |
| Dilution | ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000, Enzyme Immunoassay, Competitive ELISA |
| Applications | ELISA, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
Description
C-Peptide Monoclonal specifically detects C-Peptide in Human samples. It is validated for ELISA, Radioimmunoassay, Enzyme Immunoassay, Competitive ELISA.Specifications
| C-Peptide | |
| ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000, Enzyme Immunoassay, Competitive ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 3630 | |
| Derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant C-peptide construction. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 70000000000 | |
| ELISA, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Mouse | |
| Diabetes Research | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| INS | |
| IgG1 κ | |
| Protein A purified | |
| This is reacting only with human C-peptide. Binds to the human free C-peptide and C-peptide region in proinsulin molecules. Antigenic determinants identified by monoclnal antibodies are in the three-dimensional structure generated by N- and C- terminal regions of C-peptide separated by beta-turn at position 47-50 of proinsulin. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title