missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Peptide Antibody (2B7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2555
This item is not returnable.
View return policy
Description
C-Peptide Monoclonal specifically detects C-Peptide in Human, Bovine (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative) samples. It is validated for ELISA, Radioimmunodiffusion, Radioimmunoassay, Competitive ELISA.
Specifications
| C-Peptide | |
| Monoclonal | |
| Unconjugated | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Affinity (Kd) > 1 x 10^8. Binds to the human free C-peptide and C-peptide region in proinsulin molecules. Antigenic determinants identified by MAbs are in the three-dimensional structure generated by N- and C- terminal regions of C-peptide separated by beta-turn at position 47-50 of proinsulin.. No cross-reactivity with human, bovine, porcine and mouse/rat insulin. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| ELISA, Radioimmune Assays (RIA) | |
| 2B7 | |
| ELISA 1:100-1:2000, Radioimmunodiffusion, Radioimmunoassay 1:100-1:2000, Competitive ELISA | |
| INS | |
| Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant C-peptide construction. | |
| 0.2 mg | |
| Diabetes Research | |
| 3630 | |
| Human, Bovine (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction