missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc, p Thr58 Antibody (SR2018), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | c-Myc |
|---|---|
| Clone | SR2018 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610135
|
Novus Biologicals
NBP3-21509-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692214
|
Novus Biologicals
NBP3-21509-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Myc Monoclonal antibody specifically detects c-Myc in Human, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| c-Myc | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| A synthesized peptide derived from human phospho-c-Myc [Thr58, Ser62] (Uniprot #: P01106) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR2018 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4609 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title