Learn More
Invitrogen™ c-Myc Monoclonal Antibody (9E10), eBioscience™, Invitrogen™

Description
Description: The 9E10 antibody reacts with human c-Myc p67; the antibody was raised against a synthetic peptide corresponding to amino acids 408-439 within the carboxy terminal domain of human c-Myc. This antibody can be used to detect myc-tagged protein. The transcription factor c-Myc is a proto-oncogene that is at the focal point in cell cycle regulation, metabolism, apoptosis, differentiation, cell adhesion, and tumorigenesis. In normal cells the expression of c-Myc is tightly regulated but in human cancers c-Myc is frequently deregulated. C-Myc also plays a pivotal role in apoptosis, most notably its connections to the CD95/Fas death receptor pathway. These different biological responses to c-Myc are most likely the result of different overlapping subsets of c-Myc target genes. Applications Reported: This 9E10 antibody has been reported for use in immunoprecipitation, western blotting, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This 9E10 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using low or high pH antigen retrieval and can be used at less than or equal to 5 μg/mL. This 9E10 antibody has also been tested by western blot and can be used 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE.
Specifications
Specifications
| Antigen | c-Myc |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 9E10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | MYC |
| Gene Accession No. | P01106 |
| Gene Alias | AU016757; Avian myelocytomatosis viral (v-myc) oncogene homolog; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; mMyc; MRTL; Myc; Myc protein; Myc proto-oncogene protein; MYC proto-oncogene, bHLH transcription factor; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; myelocytomatosis viral oncogene homolog; Niard; Nird; Proto-oncogene c-Myc; RNCMYC; Transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene-like protein |
| Afficher plus |
For Research Use Only.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.