missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody (9E11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£155.00 - £456.00
Specifications
| Antigen | c-Myc |
|---|---|
| Clone | 900000000000 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:200-1:400, ELISA 1:100-1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33542232), Immunoprecipitation 2ug/mg lysate, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 2 ug/ 500 ug extract, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18099545
|
Novus Biologicals
NB200-108SS |
0.025 mL |
£155.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745673
|
Novus Biologicals
NB200-108 |
0.1 mL |
£456.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Myc Monoclonal specifically detects c-Myc in Human, Mouse, Rat, Chicken, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.Specifications
| c-Myc | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| P01106 | |
| 4609 | |
| A synthetic peptide corresponding to amino acids 408-420 (AEEQKLISEEDL) of human c-Myc Antibody (9E11), conjugated to KLH. [UniProt# P01106] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 48.8 kDa |
| 900000000000 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:200-1:400, ELISA 1:100-1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33542232), Immunoprecipitation 2ug/mg lysate, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 2 ug/ 500 ug extract, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Chicken, Yeast | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| MYC | |
| IgG2a κ | |
| Protein A or G purified | |
| Human, mouse, Saccharomyces and chicken. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title