missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody (9E11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80674
This item is not returnable.
View return policy
Description
c-Myc Monoclonal specifically detects c-Myc in Human, Mouse, Rat, Chicken, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.
Specifications
| c-Myc | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:500-1:1000, Flow Cytometry : 1:200-1:400, ELISA : 1:100-1:2000, Immunohistochemistry : 1:100, Immunoprecipitation : 2ug/mg lysate, Immunohistochemistry-Paraffin : 1:100, Immunohistochemistry-Frozen : 1:100, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 2 ug, CyTOF-ready | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| Mouse | |
| 48.8 kDa | |
| 0.1 mL | |
| Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 4609 | |
| Human, Mouse, Rat, Chicken, Yeast | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoprecipitation | |
| 9e+011 | |
| Unconjugated | |
| PBS | |
| MYC | |
| A synthetic peptide corresponding to amino acids 408-420 (AEEQKLISEEDL) of human c-Myc Antibody (9E11), conjugated to KLH. [UniProt# P01106] | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Rat reactivity reported in scientific literature (PMID: 15777849). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction