Learn More
c-Myc Antibody (2270A), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB36961J
Product Code. 18640140
  • £531.00 / 0.10mL
Description
c-Myc Monoclonal antibody specifically detects c-Myc in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF, Gene Knock-Out
TRUSTED_SUSTAINABILITY
Specifications
Specifications
c-Myc
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF, Gene Knock-Out
2270A
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated
avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
E.coli-derived recombinant human c-Myc, Arg66-Asp201, Accession # P01106
0.1 mL
Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
4609
Store at 4°C in the dark.